Assay Design



Gene Transcripts

Gene Symbol
Arrdc3
Entrez Gene ID
309945
Gene Name
arrestin domain containing 3
Gene Aliases
LRRGT00048
Chromosome Location
Chr.2: 8732910 - 8745448 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007797.1 NP_001007798.1 5 - 6 874 76
GenBank mRNA AY383703.1 - 5 - 6 874 76

View Full Product Details