Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107647.2 | NP_001101117.1 | 9 - 10 | 1225 | 77 |
XM_006231836.2 | XP_006231898.1 | 9 - 10 | 1653 | 77 | |
XM_008760671.1 | XP_008758893.1 | 8 - 9 | 3848 | 77 | |
XM_008760672.1 | XP_008758894.1 | 9 - 10 | 3700 | 77 | |
XM_008760673.1 | XP_008758895.1 | 9 - 10 | 1457 | 77 | |
GenBank mRNA | BC131851.1 | - | 9 - 10 | 1225 | 77 |