Assay Design



Gene Transcripts

Gene Symbol
Slc25a23
Entrez Gene ID
301113
Gene Name
solute carrier family 25 (mitochondrial carrier; phosphate carrier), member 23
Gene Aliases
-
Chromosome Location
Chr.9: 9988057 - 10001962 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106873.2 NP_001100343.1 6 - 7 796 66

View Full Product Details