Assay Design



Gene Transcripts

Gene Symbol
Asb6
Entrez Gene ID
296627
Gene Name
ankyrin repeat and SOCS box-containing 6
Gene Aliases
-
Chromosome Location
Chr.3: 9659707 - 9664368 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011963.1 NP_001011963.1 2 - 3 441 127
XM_006233912.2 XP_006233974.1 2 - 3 409 127
XM_008761641.1 XP_008759863.1 1 - 2 1175 127
XM_008761642.1 XP_008759864.1 3 - 4 420 127
GenBank mRNA BC079099.1 - 2 - 3 441 127

View Full Product Details