Assay Design



Gene Transcripts

Gene Symbol
Tada1
Entrez Gene ID
360874
Gene Name
transcriptional adaptor 1
Gene Aliases
RGD1306774, Tada1l
Chromosome Location
Chr.13: 84588587 - 84602542 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037980.2 NP_001033069.2 5 - 6 641 77
GenBank mRNA BC091380.1 - - 433 77
BC158713.1 - 5 - 6 641 77

View Full Product Details