Assay Design



Gene Transcripts

Gene Symbol
Txnl1
Entrez Gene ID
140922
Gene Name
thioredoxin-like 1
Gene Aliases
Txnl
Chromosome Location
Chr.18: 59056454 - 59086251 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080887.2 NP_543163.1 7 - 8 1046 93
GenBank mRNA BC098908.1 - 7 - 8 1046 93
FQ213590.1 - 7 - 8 1017 93
FQ226669.1 - 7 - 8 996 93

View Full Product Details