Assay Design



Gene Transcripts

Gene Symbol
Tnfrsf26
Entrez Gene ID
361685
Gene Name
tumor necrosis factor receptor superfamily, member 26
Gene Aliases
-
Chromosome Location
Chr.1: 216808642 - 216829366 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108511.1 NP_001101981.1 6 - 7 567 88

View Full Product Details