Assay Design



Gene Transcripts

Gene Symbol
Ctsw
Entrez Gene ID
293676
Gene Name
cathepsin W
Gene Aliases
-
Chromosome Location
Chr.1: 220844579 - 220847967 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024242.1 NP_001019413.1 8 - 9 815 67
GenBank mRNA BC093401.1 - 8 - 9 815 67
FQ235197.1 - 5 - 6 397 67

View Full Product Details