Assay Design



Gene Transcripts

Gene Symbol
Tut1
Entrez Gene ID
499314
Gene Name
terminal uridylyl transferase 1, U6 snRNA-specific
Gene Aliases
-
Chromosome Location
Chr.1: 225151462 - 225162445 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033901.1 NP_001029073.1 6 - 7 1289 120
GenBank mRNA BC104695.1 - 6 - 7 1289 120

View Full Product Details