Assay Design



Gene Transcripts

Gene Symbol
Ms4a3
Entrez Gene ID
293753
Gene Name
membrane-spanning 4-domains, subfamily A, member 3
Gene Aliases
-
Chromosome Location
Chr.1: 227979307 - 227994208 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006231111.2 XP_006231173.1 6 - 7 778 111
XM_008760278.1 XP_008758500.1 4 - 5 484 111

View Full Product Details