Assay Design



Gene Transcripts

Gene Symbol
Ric8a
Entrez Gene ID
293614
Gene Name
RIC8 guanine nucleotide exchange factor A
Gene Aliases
Ric-8A, Ric8
Chromosome Location
Chr.1: 213606599 - 213612905 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100520.1 NP_001093990.1 2 - 3 364 86
GenBank mRNA AY177754.1 - 2 - 3 124 86
BC160852.1 - 2 - 3 364 86
FQ223245.1 - - 153 86

View Full Product Details