Assay Design



Gene Transcripts

Gene Symbol
Ric8a
Entrez Gene ID
293614
Gene Name
RIC8 guanine nucleotide exchange factor A
Gene Aliases
Ric-8A, Ric8
Chromosome Location
Chr.1: 213606599 - 213612905 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100520.1 NP_001093990.1 7 - 8 1435 67
GenBank mRNA BC160852.1 - 7 - 8 1435 67
FQ223245.1 - - 1130 67

View Full Product Details