Assay Design
Gene Transcripts
- Gene Symbol
- Psbpc2
- Entrez Gene ID
-
293731
- Gene Name
- prostatic steroid-binding protein C2
- Gene Aliases
-
Psbp2
- Chromosome Location
-
Chr.1: 225690756 - 225693631 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_207593.2 |
NP_997476.2 |
1 - 2 |
85 |
69 |
GenBank mRNA |
BC099748.1 |
- |
1 - 2 |
79 |
69 |
View Full Product Details