Assay Design



Gene Transcripts

Gene Symbol
Etf1
Entrez Gene ID
307503
Gene Name
eukaryotic translation termination factor 1
Gene Aliases
-
Chromosome Location
Chr.18: 27698693 - 27725694 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008344.1 NP_001008345.1 4 - 5 502 152
GenBank mRNA BC085902.1 - 4 - 5 502 152

View Full Product Details