Assay Design



Gene Transcripts

Gene Symbol
Rit2
Entrez Gene ID
291713
Gene Name
Ras-like without CAAX 2
Gene Aliases
Rin, Rit1
Chromosome Location
Chr.18: 23700012 - 24057917 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013060.1 NP_001013078.1 2 - 3 469 67
GenBank mRNA BC091382.1 - 2 - 3 469 67
FQ213009.1 - 2 - 3 266 67

View Full Product Details