Assay Design



Gene Transcripts

Gene Symbol
Cmpk1
Entrez Gene ID
298410
Gene Name
cytidine monophosphate (UMP-CMP) kinase 1
Gene Aliases
CK, Cmpk, RGD1310116
Chromosome Location
Chr.5: 133759722 - 133786452 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025655.1 NP_001020826.1 1 - 2 260 98
GenBank mRNA BC098727.1 - 1 - 2 260 98

View Full Product Details