Assay Design



Gene Transcripts

Gene Symbol
Racgap1
Entrez Gene ID
315298
Gene Name
Rac GTPase-activating protein 1
Gene Aliases
-
Chromosome Location
Chr.7: 141277303 - 141307298 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108112.1 NP_001101582.1 3 - 4 476 81
XM_006257400.2 XP_006257462.1 3 - 4 423 81
GenBank mRNA BC166473.1 - 3 - 4 376 81

View Full Product Details