Assay Design
Gene Transcripts
- Gene Symbol
- Dhh
- Entrez Gene ID
-
84380
- Gene Name
- desert hedgehog
- Gene Aliases
-
-
- Chromosome Location
-
Chr.7: 140575288 - 140580783 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_053367.1 |
NP_445819.1 |
2 - 3 |
857 |
57 |
View Full Product Details