Assay Design
Gene Transcripts
- Gene Symbol
- Sp7
- Entrez Gene ID
-
300260
- Gene Name
- Sp7 transcription factor
- Gene Aliases
-
Osx
- Chromosome Location
-
Chr.7: 143958520 - 143967484 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_181374.2 |
NP_852039.2 |
1 - 2 |
114 |
107 |
View Full Product Details