Assay Design



Gene Transcripts

Gene Symbol
Dnajc22
Entrez Gene ID
362998
Gene Name
DnaJ (Hsp40) homolog, subfamily C, member 22
Gene Aliases
RGD1311098
Chromosome Location
Chr.7: 140781692 - 140786929 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014204.1 NP_001014226.1 1 - 2 63 64
GenBank mRNA BC086949.1 - 1 - 2 63 64

View Full Product Details