Assay Design



Gene Transcripts

Gene Symbol
Mettl7a
Entrez Gene ID
315306
Gene Name
methyltransferase like 7A
Gene Aliases
RGD1308407
Chromosome Location
Chr.7: 141973553 - 141981432 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037355.1 NP_001032432.1 1 - 2 534 87
GenBank mRNA BC105760.1 - 1 - 2 534 87

View Full Product Details