Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001109506.1 | NP_001102976.1 | 6 - 7 | 641 | 63 |
XM_006255302.2 | XP_006255364.1 | 6 - 7 | 888 | 63 | |
XM_006255303.2 | XP_006255365.1 | 6 - 7 | 687 | 63 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_008772180.1 | XP_008770402.1 | 1 - 1 | 612 | 63 |