Assay Design



Gene Transcripts

Gene Symbol
Sntb2
Entrez Gene ID
689421
Gene Name
syntrophin, beta 2
Gene Aliases
-
Chromosome Location
Chr.19: 39126589 - 39209152 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168674.1 NP_001162145.1 3 - 4 1036 70

View Full Product Details