Assay Design



Gene Transcripts

Gene Symbol
Vat1l
Entrez Gene ID
361414
Gene Name
vesicle amine transport 1-like
Gene Aliases
-
Chromosome Location
Chr.19: 46525852 - 46693838 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003751883.3 XP_003751931.1 8 - 9 1310 60
GenBank mRNA FQ211768.1 - 8 - 9 1243 60

View Full Product Details