Assay Design



Gene Transcripts

Gene Symbol
Wnt2b
Entrez Gene ID
116466
Gene Name
wingless-type MMTV integration site family, member 2B
Gene Aliases
-
Chromosome Location
Chr.2: 207440886 - 207455258 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191848.1 NP_001178777.1 3 - 4 676 70
GenBank mRNA AF204873.1 - 2 - 3 436 70

View Full Product Details