Assay Design



Gene Transcripts

Gene Symbol
Slc22a15
Entrez Gene ID
310732
Gene Name
solute carrier family 22, member 15
Gene Aliases
-
Chromosome Location
Chr.2: 204281638 - 204344026 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107707.1 NP_001101177.1 2 - 3 358 96
XM_006233050.2 XP_006233112.1 1 - 2 2752 96
GenBank mRNA FQ211559.1 - 1 - 2 404 96

View Full Product Details