Assay Design



Gene Transcripts

Gene Symbol
Slc22a15
Entrez Gene ID
310732
Gene Name
solute carrier family 22, member 15
Gene Aliases
-
Chromosome Location
Chr.2: 204281638 - 204344026 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107707.1 NP_001101177.1 5 - 6 866 87
XM_006233050.2 XP_006233112.1 4 - 5 3260 87

View Full Product Details