Assay Design



Gene Transcripts

Gene Symbol
Hibadh
Entrez Gene ID
63938
Gene Name
3-hydroxyisobutyrate dehydrogenase
Gene Aliases
-
Chromosome Location
Chr.4: 82604369 - 82702429 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022243.1 NP_071579.1 4 - 5 510 93
GenBank mRNA BC127442.1 - 4 - 5 496 93
J04628.1 - 4 - 5 511 93

View Full Product Details