Assay Design



Gene Transcripts

Gene Symbol
Cd47
Entrez Gene ID
29364
Gene Name
Cd47 molecule
Gene Aliases
Itgp
Chromosome Location
Chr.11: 53511485 - 53575754 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019195.2 NP_062068.1 8 - 9 1019 107
XM_006248264.2 XP_006248326.1 9 - 10 1712 107
XM_006248265.2 XP_006248327.1 10 - 11 1757 107
GenBank mRNA BC085740.1 - 8 - 9 1019 107
FQ226437.1 - 2 - 3 131 107
FQ232147.1 - 4 - 5 273 107
FQ232168.1 - 3 - 4 211 107
FQ232318.1 - 7 - 8 642 107
FQ233021.1 - 2 - 3 131 107
FQ233572.1 - 3 - 4 191 107

View Full Product Details