Assay Design
Gene Transcripts
- Gene Symbol
- Crybg3
- Entrez Gene ID
-
288204
- Gene Name
- beta-gamma crystallin domain containing 3
- Gene Aliases
-
RGD1305628
- Chromosome Location
-
Chr.11: 42895607 - 43006568 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_213639.8 |
XP_213639.6 |
19 - 20 |
8611 |
58 |
View Full Product Details