Assay Design



Gene Transcripts

Gene Symbol
C8b
Entrez Gene ID
313421
Gene Name
complement component 8, beta polypeptide
Gene Aliases
-
Chromosome Location
Chr.5: 124298271 - 124338055 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191759.1 NP_001178688.1 5 - 6 660 146
XM_006238495.2 XP_006238557.1 6 - 7 980 146
GenBank mRNA BC090023.1 - 2 - 3 185 146

View Full Product Details