Assay Design



Gene Transcripts

Gene Symbol
Zfp36
Entrez Gene ID
79426
Gene Name
zinc finger protein 36
Gene Aliases
Tis11
Chromosome Location
Chr.1: 85380088 - 85382565 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133290.3 NP_579824.2 1 - 2 77 59
GenBank mRNA BC060308.1 - 1 - 2 81 59
X63369.1 - 1 - 2 62 59

View Full Product Details