Assay Design



Gene Transcripts

Gene Symbol
Lgi4
Entrez Gene ID
361549
Gene Name
leucine-rich repeat LGI family, member 4
Gene Aliases
-
Chromosome Location
Chr.1: 89491589 - 89501912 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199499.1 NP_955793.1 2 - 3 448 88
XM_008759170.1 XP_008757392.1 2 - 3 162 88
GenBank mRNA BC064659.1 - 2 - 3 448 88

View Full Product Details