Assay Design



Gene Transcripts

Gene Symbol
Kirrel2
Entrez Gene ID
100359836
Gene Name
kin of IRRE like 2 (Drosophila)
Gene Aliases
-
Chromosome Location
Chr.1: 88909838 - 88920834 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271398.1 NP_001258327.1 8 - 9 1713 78
XM_008759099.1 XP_008757321.1 9 - 10 1669 78

View Full Product Details