Assay Design



Gene Transcripts

Gene Symbol
Siglec10
Entrez Gene ID
292844
Gene Name
sialic acid binding Ig-like lectin 10
Gene Aliases
Siglecg
Chromosome Location
Chr.1: 98512663 - 98521740 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106250.1 NP_001099720.1 5 - 6 1348 62
XM_006228948.2 XP_006229010.1 8 - 9 1507 62
GenBank mRNA BC168854.1 - 3 - 4 581 62

View Full Product Details