Assay Design



Gene Transcripts

Gene Symbol
Nfkbid
Entrez Gene ID
308496
Gene Name
nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, delta
Gene Aliases
RGD1308055, Ta-nfkbh
Chromosome Location
Chr.1: 88883817 - 88892543 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003748825.3 XP_003748873.1 9 - 10 1454 59
XM_006228804.2 XP_006228866.1 8 - 9 1940 59

View Full Product Details