Assay Design



Gene Transcripts

Gene Symbol
Stoml2
Entrez Gene ID
298203
Gene Name
stomatin (Epb7.2)-like 2
Gene Aliases
-
Chromosome Location
Chr.5: 58472561 - 58476158 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031646.1 NP_001026816.1 5 - 6 450 63
GenBank mRNA BC099164.1 - 5 - 6 450 63

View Full Product Details