Assay Design



Gene Transcripts

Gene Symbol
Car9
Entrez Gene ID
313495
Gene Name
carbonic anhydrase 9
Gene Aliases
-
Chromosome Location
Chr.5: 59008277 - 59015535 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107956.1 NP_001101426.1 7 - 8 1051 99
XM_008763676.1 XP_008761898.1 7 - 8 1707 99
GenBank mRNA EF187254.1 - 7 - 8 1053 99

View Full Product Details