Assay Design



Gene Transcripts

Gene Symbol
Espnl
Entrez Gene ID
301606
Gene Name
espin-like
Gene Aliases
RGD1562432
Chromosome Location
Chr.9: 98459641 - 98484930 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191812.1 NP_001178741.1 1 - 2 295 144
XM_006245376.2 XP_006245438.1 1 - 2 831 144

View Full Product Details