Assay Design



Gene Transcripts

Gene Symbol
Arl5b
Entrez Gene ID
364788
Gene Name
ADP-ribosylation factor-like 5B
Gene Aliases
Arl8
Chromosome Location
Chr.17: 82065910 - 82092693 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015031.1 NP_001015031.1 1 - 2 193 75
XM_006254312.2 XP_006254374.1 1 - 2 295 75
GenBank mRNA BC091260.1 - 1 - 2 193 75

View Full Product Details