Assay Design



Gene Transcripts

Gene Symbol
Trdmt1
Entrez Gene ID
291324
Gene Name
tRNA aspartic acid methyltransferase 1
Gene Aliases
Dnmt2
Chromosome Location
Chr.17: 80826749 - 80859585 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031643.1 NP_001026813.1 9 - 10 979 119
GenBank mRNA BC098700.1 - 9 - 10 979 119

View Full Product Details