Assay Design



Gene Transcripts

Gene Symbol
Prr20e
Entrez Gene ID
685621
Gene Name
proline rich 20E
Gene Aliases
-
Chromosome Location
Chr.4: 121045875 - 121052943 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008763118.1 XP_008761340.1 2 - 3 116 66

View Full Product Details