Assay Design



Gene Transcripts

Gene Symbol
Tmprss11e
Entrez Gene ID
305265
Gene Name
transmembrane protease, serine 11e
Gene Aliases
RGD1561698
Chromosome Location
Chr.14: 23069213 - 23113258 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008770102.1 XP_008768324.1 2 - 3 123 78

View Full Product Details