Assay Design



Gene Transcripts

Gene Symbol
Yipf7
Entrez Gene ID
364147
Gene Name
Yip1 domain family, member 7
Gene Aliases
RGD1310766
Chromosome Location
Chr.14: 62567841 - 62609508 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108861.1 NP_001102331.1 1 - 2 47 153
XM_006251039.2 XP_006251101.1 2 - 3 111 153

View Full Product Details