Assay Design
Gene Transcripts
- Gene Symbol
- Tlr6
- Entrez Gene ID
-
305353
- Gene Name
- toll-like receptor 6
- Gene Aliases
-
-
- Chromosome Location
-
Chr.14: 45024351 - 45041188 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_207604.1 |
NP_997487.1 |
1 - 2 |
217 |
86 |
GenBank mRNA |
BC062390.1 |
- |
1 - 2 |
217 |
86 |
View Full Product Details