Assay Design



Gene Transcripts

Gene Symbol
Fuca2
Entrez Gene ID
292485
Gene Name
fucosidase, alpha-L- 2, plasma
Gene Aliases
-
Chromosome Location
Chr.1: 7658919 - 7673663 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004218.1 NP_001004218.1 6 - 7 1324 96
GenBank mRNA BC078933.1 - 6 - 7 1324 96

View Full Product Details