Assay Design



Gene Transcripts

Gene Symbol
Ankdd1a
Entrez Gene ID
100362785
Gene Name
ankyrin repeat and death domain containing 1A
Gene Aliases
-
Chromosome Location
Chr.8: 71034630 - 71057264 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006243300.2 XP_006243362.1 10 - 11 2233 103

View Full Product Details