Assay Design



Gene Transcripts

Gene Symbol
Bco2
Entrez Gene ID
315644
Gene Name
beta-carotene oxygenase 2
Gene Aliases
Ab2-079, Bcdo2
Chromosome Location
Chr.8: 54965577 - 54990604 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127712.1 NP_001121184.1 6 - 7 824 101
GenBank mRNA BC133725.1 - 6 - 7 824 101
DQ083174.1 - 4 - 5 488 101

View Full Product Details