Assay Design



Gene Transcripts

Gene Symbol
Arhgap20
Entrez Gene ID
367085
Gene Name
Rho GTPase activating protein 20
Gene Aliases
RARhoGAP
Chromosome Location
Chr.8: 56179586 - 56264343 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213629.1 NP_998794.1 14 - 15 1799 63
XM_006243047.2 XP_006243109.1 14 - 15 1810 63
GenBank mRNA AY501475.1 - 14 - 15 1799 63

View Full Product Details