Assay Design



Gene Transcripts

Gene Symbol
Trappc5
Entrez Gene ID
363858
Gene Name
trafficking protein particle complex 5
Gene Aliases
RGD1563971
Chromosome Location
Chr.12: 2228670 - 2232973 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108850.2 NP_001102320.1 1 - 2 63 70
GenBank mRNA BC158788.1 - 1 - 2 63 70
FQ227574.1 - 1 - 2 78 70

View Full Product Details